Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Osteostat (from E. coli)
Human Recombinant Osteostat (from E. coli)
Catalog # 200068-174
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Osteostat (from E. coli)
Catalog # 200068-174
Supplier:  BioVendor
Supplier Number:  RD172111100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    TNFS18|AITR Ligand
  • Protein/Peptide Name:
    Osteostat
  • Sequence:
    MRGSHHHHHH GMASTAKEPC MAKFGPLPSK WQMASSEPPC VNKVSDWKLE ILQNGLYLIY GQVAPNANYN DVAPFEVRLY KNKDMIQTLT NKSKIQNVGG TYELHVGDTI DLIFNSEHQV LKNNTYWGII LLANPQFIS
  • Cat. No.:
    200068-174
  • Supplier no.:
    RD172111100

Specifications

About this item

Osteostat is the cytokine that binds to TNFRSF18/AITR/GITR and is important for interactions between activated T-lymphocytes and endothelial cells and may modulate T-lymphocyte survival in peripheral tissues. Osteostat is expressed at high levels in the small intestine, ovary, testis, kidney and endothelial cells after stimulation by lipopolysaccharides.

Sequence: MRGSHHHHHH GMASTAKEPC MAKFGPLPSK WQMASSEPPC VNKVSDWKLE ILQNGLYLIY GQVAPNANYN DVAPFEVRLY KNKDMIQTLT NKSKIQNVGG TYELHVGDTI DLIFNSEHQV LKNNTYWGII LLANPQFIS

Other names: TNFS18, AITR Ligand

Ordering information: For research use only.