Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Protein Synonyms:ZSIG51|Thyrostimulin alpha subunit|GPHB5|Glycoprotein hormone beta 5|Putative secreted protein Zsig51|Thyrostimulin beta subunit|Glycoprotein hormone alpha-2|GPA2|GPHA2
- Protein/Peptide Name:Thyrostimulin
- Sequence:MRGSHHHHHH GMASQEAVIP GCHLHPFNVT VRSDRQGTCQ GSHVAQACVG HCESSAFPSR YSVLVASGYR HNITSVSQCC TISGLKKVKV QLQCVGSRRE ELEIFTARAC QCDMCRLSRY
- Cat. No.:200068-170
- Supplier no.:RD172110100
Human thyrostimulin ranks among the glycoprotein hormone family. These hormones consist of two subunits, the common alpha- and specific beta-subunits, which associate noncovalently to form a heterodimer. The alpha-subunit combines with four distinct beta-subunits giving rise to four biologically active hormones in human: FSH, LH, TSH, and CG. FSH, LH, and TSH, mainly expressed in the anterior pituitary, are essential for coordinated endocrine regulation in the hypothalamus- pituitary axis and show to activate specific G protein–coupled receptors in the thyroid (TSH receptor) and gonads (LH and FSH receptors), respectively.
Sequence: MRGSHHHHHH GMASQEAVIP GCHLHPFNVT VRSDRQGTCQ GSHVAQACVG HCESSAFPSR YSVLVASGYR HNITSVSQCC TISGLKKVKV QLQCVGSRRE ELEIFTARAC QCDMCRLSRY
Other names: Thyrostimulin alpha subunit, GPHA2, Glycoprotein hormone beta 5, GPHB5, Thyrostimulin beta subunit, Glycoprotein hormone alpha-2, Putative secreted protein Zsig51, GPA2, ZSIG51