Order Entry
Puerto Rico
ContactUsLinkComponent
Human Regenerating-1 (fromE. coli)
Human Regenerating-1 (fromE. coli)
Catalog # 101215-294
Supplier:  BioVendor
CAS Number:  
undefined
Human Regenerating-1 (fromE. coli)
Catalog # 101215-294
Supplier:  BioVendor
Supplier Number:  RD172084100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    Regenerating protein I beta|REG 1 beta|Regenerating islet-derived protein 1-beta|Pancreatic stone protein 2|REG1B|PSPS2|REG-1-beta|PSP-2|Lithostathine-1-beta|REGL
  • Protein/Peptide Name:
    Regenerating-1
  • Molecular Weight:
    17.8 kDa
  • Sequence:
    MKHHHHHHAS HMQESQTELP NPRISCPEGT NAYRSYCYYF NEDPETWVDA DLYCQNMNSG NLVSVLTQAE GAFVASLIKE SSTDDSNVWI GLHDPKKNRR WHWSSGSLVS YKSWDTGSPS SANAGYCASL TSCSGFKKWK DESCEKKFSF VCKFKN
  • Cat. No.:
    101215-294
  • Supplier no.:
    RD172084100

Specifications

About this item

Total 156 AA. MW: 17.8 kDa (calculated). UniProtKB acc.no. P48304. N-Terminal His-tag 12 AA

Sequence: MKHHHHHHAS HMQESQTELP NPRISCPEGT NAYRSYCYYF NEDPETWVDA DLYCQNMNSG NLVSVLTQAE GAFVASLIKE SSTDDSNVWI GLHDPKKNRR WHWSSGSLVS YKSWDTGSPS SANAGYCASL TSCSGFKKWK DESCEKKFSF VCKFKN

Other names: REG 1 beta, Lithostathine-1-beta, Regenerating islet-derived protein 1-beta, REG-1-beta, Regenerating protein I beta, Pancreatic stone protein 2, PSP-2, REG1B, PSPS2, REGL

Ordering information: For research use only.