Order Entry
Puerto Rico
ContactUsLinkComponent
Human Proguanylin (from E. coli)
Human Proguanylin (from E. coli)
Catalog # 101215-258
Supplier:  BioVendor
CAS Number:  
undefined
Human Proguanylin (from E. coli)
Catalog # 101215-258
Supplier:  BioVendor
Supplier Number:  RD172046100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    GUCA2|Guanylate cyclase-activating protein 1|Guanylate cyclase-activating protein I|Guanylate cyclase activator 2A|GCAP-I|GUCA2A
  • Protein/Peptide Name:
    Proguanylin
  • Molecular Weight:
    11,5 kDa
  • Sequence:
    MKHHHHHHAS VTVQDGNFSF SLESVKKLKD LQEPQEPRVG KLRNFAPIPG EPVVPILCSN PNFPEELKPL CKEPNAQEIL QRLEEIAEDP GTCEICAYCTGC
  • Cat. No.:
    101215-258
  • Supplier no.:
    RD172046100

Specifications

About this item

Guanylin is thought to modulate intestinal water/electrolyte transport in a paracrine mode reported the nucleotide sequence of the gene, the characteristics of its circulating molecular form, and its localization in enterochromaffin cells of the gut. The gene, approximately 2.6 kb in size, consists of 3 exons interrupted by 2 introns. The hormonal form of guanylin is a 94-amino acid peptide with a molecular mass of 10.3 kDa. Guanylin is synthesized by gut enterochromaffin cells as a prohormone of 115 amino acids and is processed to the molecular form of 94 amino acids circulating in the blood. Total 104 AA. MW: 11,5 kDa (calculated). UniProtKB acc.no. Q02747 (Val22-Cys115). N-terminal His-tag (10 extra AA). Protein identity confirmed by LC-MS/MS.

Sequence: MKHHHHHHAS VTVQDGNFSF SLESVKKLKD LQEPQEPRVG KLRNFAPIPG EPVVPILCSN PNFPEELKPL CKEPNAQEIL QRLEEIAEDP GTCEICAYAA CTGC

Other names: Guanylate cyclase activator 2A, Guanylate cyclase-activating protein 1, Guanylate cyclase-activating protein I, GCAP-I, GUCA2A, GUCA2

Ordering information: For research use only.