Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.025 mg
- Storage Conditions:Store lyophilized protein at –80 °C. Lyophilized protein remains stable until the expiry date when stored at –80 °C. Aliquot reconstituted protein to avoid repeated freezing/thawing cycles and store at –80 °C for long term storage. Reconstituted protein can be stored at 4 °C for a week
- Endotoxin Content:<1.0 EU/µg
- Reconstitution Instructions:Add deionized water to prepare a working stock solution of approximately 0.25 mg/ml and let the lyophilized pellet dissolve completely.
- Protein Synonyms:NEFL|NFL|NF-L|68 kDa neurofilament protein|Neurofilament triplet L protein|NF68
- Protein/Peptide Name:Neurofilament light polypeptide Human E. coli
- Purity:>90%
- Molecular Weight:62.5 kDa (calculated)
- Sequence:MKHHHHHHASSFSYEPYYSTSYKRRYVETPRVHISSVRSGYSTARSAYSSYSAPVSSSLSVRRSYSSSSGSLMPSLENLDLSQVAAISNDLKSIRTQEKAQLQDLNDRFASFIERVHELEQQNKVLEAELLVLRQKHSEPSRFRALYEQEIRDLRLAAEDATNEKQALQGEREGLEETLRNLQARYEEEVLSREDAEGRLMEARKGADEAALARAELEKRIDSLMDEISFLKKVHEEEIAELQAQIQYAQISVEMDVTKPDLSAALKDIRAQYEKLAAKNMQNAEEWFKSRFTVLTESAAKNTDAVRAAKDEVSESRRLLKAKTLEIEACRGMNEALEKQLQELEDKQNADISAMQDTINKLENELRTTKSEMARYLKEYQDLLNVKMALDIEIAAYRKLLEGEETRLSFTSVGSITSGYSQSSQVFGRSAYGGLQTSSYLMSTRSFPSYYTSHVQEEQIEVEETIEAAKAEEAKDEPPSEGEAEEEEKDKEEAEEEEAAEEEEAAKEESEEAKEEEEGGEGEEGEETKEAEEEEKKVEGAGEEQAAKKKD
- Endotoxin Level:<1.0 EU/µg
- Formulation:Filtered (0.4 μm) and lyophilized from 0.5 mg/ml solution in 15 mM Tris, 85 mM Glycine, pH 8.6
- Shipping Temperature:At ambient temperature
- Tested Applications:Western blotting, ELISA
- Cat. No.:101189-770
- Supplier no.:RD172009100-H
Total 551 AA. MW: 62.5 kDa (calculated). UniProtKB acc.no. P07196 (Ser2-Asp543). N-terminal His-tag (9 extra AA). Protein identity confirmed by LC-MS/MS.
- Quality control test
- BCA to determine quantity of the protein
- SDS PAGE to determine purity of the protein
- LAL to determine quantity of endotoxin