QuantiTect SYBR Green PCR Kit, Qiagen
Supplier: Qiagen
Certificates
About this item
QuantiTect SYBR® Green PCR kits for real-time PCR and two-step RT-PCR using SYBR® Green.
- High PCR specificity with integrated hot start
- Reliable quantification of low-abundance transcripts
- Accurate quantification
- Available with or without uracil-N-glycosylase (UNG)
- No need to optimize reaction and cycling conditions
Includes 1mL 2x QuantiTect SYBR® Green PCR Master Mix and 2mL RNase-free water.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.