QuantiTect Multiplex PCR Kit , Qiagen
: Qiagen
QuantiTect Multiplex PCR kits for multiplex, real-time PCR and RT-PCR using sequence-specific probes.
- Multiplex analysis with no need for optimization
- Sensitive detection of as few as 10 copies of each target
- Reliable quantification of target and reference genes
- Detection of up to 5 targets in the same tube
Includes 1mL 2x QuantiTect Multiplex PCR Master Mix (with ROX dye), and 2mL RNase-free water.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.