ContactUsLinkComponent
HydroLogix HLT 5000 Pipette Tips, Molecular BioProducts
HydroLogix HLT 5000 Pipette Tips, Molecular BioProducts
  72830-734
   Low Manufacturing Impact
HydroLogix HLT 5000 Pipette Tips, Molecular BioProducts
  72830-734
 :  3897-11
   Low Manufacturing Impact
  • Volume:
    5000 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Tip color:
    Clear
  • Packed:
    Bulk, 250 Tips/bag
  • Cat. No.:
    72830-734
  • Tip style:
    Standard Tips
  • Supplier no.:
    3897-11
These non-filtered tips are made of polypropylene.
  • Clear
  • MBP clear HydroLogix tip
  • Non-sterile