FastRNA™, Spin Kit, MP Biomedicals
The FastRNA™ Spin Kit isolates both large and small RNA species without the use of phenol or reducing agents.
- Spin Kit for Microbes quickly and efficiently isolates high-quality, total RNA from tough-to-lyse bacterial cell cultures
- Spin Kit for Yeast quickly and efficiently isolates high-quality, total RNA from tough-to-lyse yeast strains, fungi, and algae
The FastRNA™ SPIN Kit for Microbes quickly and efficiently isolates high-quality, total RNA from tough-to-lyse bacterial cell cultures and other microbial sources in approximately 15 minutes using a zirconium silicate Lysing Matrix (0.1mm) to lyse bacteria and SPIN columns for the purification process.
The FastRNA™ SPIN Kit for Yeast quickly and efficiently isolates high-quality, total RNA from tough-to-lyse yeast strains, fungi, and algae in approximately 15 minutes using a specialized Lysing Matrix (Yttria-stabilized zirconium oxide, 0.5mm) for cell lysis and SPIN columns for the purification process.
- Sample type:Bacteria
- Storage Conditions:All FastRNA™ SPIN Kit for Microbes reagents are shipped ambiently. FastRNA™ SPIN Kit for Microbes reagents are stable at room temperature. (15-30 °C)
- Instrument Compatibility:FastPrep-24; FastPrep-96
- Description:FastRNA™ Spin Kit for Microbes
Frequently Bought Together