Septa for 2.0 ml Big Mouth Step Vials, Restek
Catalog # 10854-010
Supplier: Restek
Specifications
- Description:Septa only
- Cat. No.:10854-010
- Supplier no.:21148
Specifications
About this item
These replacement septa are extremely economical accessories for use with big mouth step vials.
- Screw-thread vial compatibility
- Economical
- Red PTFE/Silicone
Extra-wide openings decrease the number of broken needles with a larger target area. These septa necessarily help maintain a seal during autosampler use.