Installation Kit for Packed Columns, Restek
Catalog # 10855-882
Supplier: Restek
Specifications
- Description:Packed Column Installation Kit
- Cat. No.:10855-882
- Supplier no.:21067
Specifications
About this item
For use with 2 mm ID columns in valve applications.
Installation kits include 1/8" stainless steel nut (1), stainless steel Valco nut (1), 1/8" Vespel/graphite ferrule (1), stainless steel Valco ferrule (1), 1/8" stainless steel front ferrule (1), and 1/8" stainless steel back ferrule (1).
Delivery: Installation kits include 1/8" stainless steel nut (1), stainless steel Valco nut (1), 1/8" Vespel/graphite ferrule (1), stainless steel Valco ferrule (1), 1/8" stainless steel front ferrule (1), and 1/8" stainless steel back ferrule (1).