FID Maintenance Kits for Agilent GCs, Restek
Supplier: Restek
Certificates
About this item
For use with Agilent 6850, 6890, and 7890 GCs.
- Kit includes the most common consumable GC supplies and tools
- Parts list makes reordering easy
- All parts meet or exceed performance of instrument manufacturer's parts