Uniliner® Inlet Liner with Top Taper/On-Column for Agilent ¼" Packed Injection Ports, Restek
Catalog # 75779-296
Supplier: Restek
Specifications
- Ext.Ø:5 mm
- Volume:0.539 ml
- Description:Uniliner Liner (large buffer volume chamber- 85 mm long, for injections ≤4 µl)
- Material:Borosilicate glass
- Cat. No.:75779-296
- For use with:AGLS
- I.D.×L:3.5×85 mm
- Treatment:Siltek
- Supplier no.:20305-214.5
Specifications
About this item
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.