ContactUsLinkComponent
Gold-Plated Liner Cap for Split/Splitless Injector, Restek
Gold-Plated Liner Cap for Split/Splitless Injector, Restek
Catalog # 10855-936
Supplier:  Restek
undefined
Gold-Plated Liner Cap for Split/Splitless Injector, Restek
Catalog # 10855-936
Supplier:  Restek
Supplier Number:  22089
Specifications
  • Description:
    Gold-Plated Liner Cap
  • Cat. No.:
    10855-936
  • Supplier no.:
    22089
Specifications
About this item
For use with split/splitless injectors on Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL.
  • Gold-plated for inertness