ContactUsLinkComponent
FID Jet Removal Tool for Agilent 5890/6850/6890/7890 FIDs, Restek
FID Jet Removal Tool for Agilent 5890/6850/6890/7890 FIDs, Restek
Catalog # 10803-284
Supplier:  Restek
undefined
FID Jet Removal Tool for Agilent 5890/6850/6890/7890 FIDs, Restek
Catalog # 10803-284
Supplier:  Restek
Supplier Number:  22328
Specifications
  • Description:
    FID Jet Removal Tool
  • Cat. No.:
    10803-284
  • For use with:
    for Agilent 5890/6850/6890/7890 GCs
  • Supplier no.:
    22328
Specifications
About this item
FID jet removal tools securely grip jet in socket for easy removal or installation.
  • Unique, ergonomic handle that is easy to hold
  • For use with Agilent 5890, 6850, 6890, and 7890 FIDs