EPC Test Kit for Agilent GCs, Restek
Catalog # 10856-136
Supplier: Restek
Specifications
- Description:EPC Test Kit
- Cat. No.:10856-136
- For use with:for Agilent 6890 GCs
- Supplier no.:24323
Specifications
About this item
EPC test kits are for use with Agilent 6890 GCs.
Kit includes three O-rings, two plugs, one mounting screw, and one test block.
Ordering information: Similar to Agilent part # G1530-60960, but not exact equivalent. Kits differ in parts.