ContactUsLinkComponent
Septum Nut for Shimadzu GCs, Restek
Septum Nut for Shimadzu GCs, Restek
Catalog # 10855-926
Supplier:  Restek
undefined
Septum Nut for Shimadzu GCs, Restek
Catalog # 10855-926
Supplier:  Restek
Supplier Number:  22079
Specifications
  • Description:
    Septum Nut
  • Cat. No.:
    10855-926
  • For use with:
    for Shimadzu 17A, 2010, and 2014 GCs
  • Supplier no.:
    22079
Specifications
About this item
Made of clear anodized aluminum and high-quality stainless steel. One-piece design for ease of installation and removal.