Order Entry
Puerto Rico
ContactUsLinkComponent
 
Silicone O-Rings for PerkinElmer Auto SYS XL or Clarus With CAP Injector, Restek
  10057-896
 :  Restek
undefined
Silicone O-Rings for PerkinElmer Auto SYS XL or Clarus With CAP Injector, Restek
  10057-896
 :  Restek
 :  20262

 

  • Description:
    O-Rings
  • Material:
    Silicone
  • Cat. No.:
    10057-896
  • For use with:
    for PerkinElmer Auto SYS XL or Clarus w/CAP Injector
  • Supplier no.:
    20262

 

 

O-rings are made of silicone.

  • Fit 6.2 mm OD liners
  • Silicone fabrication
  • Max operating temperature of 250°C

For use with PerkinElmer Auto SYS XL or Clarus with CAP injector.