Injection Port Weldments for Agilent GCs with Tekmar Purge and Trap Systems, Restek
Supplier: Restek
Certificates
About this item
Easily attach your purge-and-trap system with pre-installed low-dead-volume fittings.
For use with Agilent GCs with Tekmar purge-and-trap systems.