ContactUsLinkComponent
Rough Pump Oil #19 for MSD Pumps, Oil Vacuum Pump, Restek
Rough Pump Oil #19 for MSD Pumps, Oil Vacuum Pump, Restek
Catalog # 10057-916
Supplier:  Restek
undefined
Rough Pump Oil #19 for MSD Pumps, Oil Vacuum Pump, Restek
Catalog # 10057-916
Supplier:  Restek
Supplier Number:  22687
Specifications
  • Description:
    Rough Pump Oil for MSD Pumps
  • Cat. No.:
    10057-916
  • Supplier no.:
    22687
Specifications
About this item
Rough pump oil is formulated from crude oil stocks known for their durability and line-lubricating qualities.
  • Long-lasting lubrication
Use in Agilent 5973/5972/5971 and GCD mass spec systems, or in other manufacturers' MSD systems that require rough pump oil.
Under average use, the oil in the foreline rough pump should be replaced every six months.