PTFE Tip, Gas-Tight Syringe Replacement Needles for Removable Needle Syringes, SGE, Restek
10059-292
: Restek
- Description:PTFE-Tip, Gas-Tight Syringe Replacement Needles
- Length:50.0 mm (1.97")
- Cat. No.:10059-292
- Point style:2
- Gauge:26
- Supplier no.:24802
For use with removable needle syringes.
- Stainless steel needle
- Nickel-plated brass hub
- PTFE-tipped
- Gas-tight