Rethreading Tool for Agilent Split/Splitless Injection Ports, Restek
Catalog # 10852-840
Supplier: Restek
Specifications
- Description:Rethreading Tool
- Cat. No.:10852-840
- Supplier no.:23018
Specifications
About this item
Rethreading tools repair worn or damaged threads in injection ports, fittings, etc.
- For 1/4" compression fittings (Agilent split/splitless injection ports)
Worn and damaged threads can allow oxygen into the system—compromising analytical results and destroying columns. The built-in guide helps prevent cross-threading.