ContactUsLinkComponent
Inlet Liner Packing Tool, Restek
Inlet Liner Packing Tool, Restek
Catalog # 10803-214
Supplier:  Restek
undefined
Inlet Liner Packing Tool, Restek
Catalog # 10803-214
Supplier:  Restek
Supplier Number:  20339
Specifications
  • Description:
    Inlet Liner Packing Tool
  • Cat. No.:
    10803-214
  • Supplier no.:
    20339
Specifications
About this item
Recommended for inlet liners with an ID ≥2 mm.
  • Accurate to a specific, measured depth
  • Can be used with all manufacturers' liners