Universal 'Y' Press-Tight® Connectors, Restek
: Restek
Universal "Y" Press-Tight® connectors feature a split column flow to two detectors.
- Split column flow to two detectors
- Deactivated fused quartz helps eliminate activity for complete inertness along the GC flow path
- Universal fit: use with 0.1–0.53 mm ID Restek columns (0.33–0.74 mm OD)
Optional Siltek® deactivation is ideal for organochlorine pesticides analysis.