CarboFrit® Puller/Inserter Tool, Restek
CarboFrit Puller/Inserter Tool
Catalog # RK21642
Supplier: Restek
Supplier #: 21642
Jump to:
Product Details & Documents
Hook end for removing CarboFrit® inserts.
- Bent end (90°) for inserting CarboFrit® inserts
- Slender design
- Easy to maneuver
Insert and remove CarboFrit® inserts easily.
Specifications
- Description:CarboFrit Puller/Inserter Tool