Tubing Cutter, Restek
Catalog # RK20192
Supplier: Restek
Specifications
- Description:Tubing Cutter
- Cat. No.:RK20192
- Cutting capacity:0.16 cm (¹⁄₁₆")
- Supplier no.:20192
Specifications
About this item
Produces square, smooth cuts in tubing.
- Eliminates tubing distortion
- Replaceable cutting wheel
Cuts hard or soft copper, aluminum, brass, stainless steel, Monel® alloy, or titanium. Efficient tubing cutter consistently cut lengths of tubing.