Order Entry
Puerto Rico
ContactUsLinkComponent
Cyclo Vu-Tight DI Liners, Restek
Cyclo Vu-Tight DI Liners, Restek
Catalog # RK20788
Supplier:  Restek
Cyclo Vu-Tight DI Liners, Restek
Catalog # RK20788
Supplier:  Restek
Supplier Number:  20788

Specifications

  • Ext.Ø:
    1/4"
  • Description:
    Cyclo Vu-Tight DI Liner
  • Cat. No.:
    RK20788
  • For use with:
    Capillary Columns (O-Ø 0.5 - 0.8 mm)
  • Column I.D.:
    0.32 - 0.53 mm
  • Supplier no.:
    20788

Specifications

About this item

Visually observe the Press-Tight® connection between the column end and liner.

  • Slotted top prevents obstruction of carrier gas flow
  • Two designs available
  • Operate in the direct injection mode

Ideal for dirty samples. Spiral bore prevents nonvolatile residue from contaminating the column.