Order Entry
Puerto Rico
ContactUsLinkComponent
FID Gauge Pack for Agilent 5890 GCs, Restek
FID Gauge Pack for Agilent 5890 GCs, Restek
Catalog # 10855-924
Supplier:  Restek
undefined
FID Gauge Pack for Agilent 5890 GCs, Restek
Catalog # 10855-924
Supplier:  Restek
Supplier Number:  22071

Specifications

  • Description:
    FID Gauge Pack
  • Cat. No.:
    10855-924
  • For use with:
    for Agilent 5890 GCs
  • Supplier no.:
    22071

Specifications

About this item

The FID gauge pack includes pressure regulators and gauges for air and hydrogen that are compatible with Agilent 5890 gas chromatographs.

  • 1/8-inch bulkhead allows easy hookup to instrument
  • Rated for inlet pressures to 250 psi (1,724 kPa)
  • Rated for outlet pressures of 0 to 60 psi (0–414 kPa)

For use with Agilent 5890 GCs.