Air Diverter for Agilent GCs, Restek
RK22076
: Restek
- Description:Air Diverter
- Cat. No.:RK22076
- For use with:for Agilent 5890/6890/7890 GCs
- Supplier no.:22076
For use with Agilent 5890/6890/7890 GCs.
- Divert GC exhaust heat up and away from the lab bench
- Meets or exceeds original manufacturer's performance
- Easy to install-no tools required
- Compatible with 4" flexible metal dryer hose