FID/Injector Cleaning Kit, Restek
RK20120
: Restek
- Description:FID/Injector Cleaning Kit
- Cat. No.:RK20120
- Supplier no.:20120
The FID/Injector cleaning kit is compatible with any injection port, allowing universal applicability.
- Stainless steel cleaning tools
: Kits include: Nylon tube brushes; pipe cleaner; stainless steel tube brushes; stainless steel surface brush; stainless steel jet reamers; and emery cloth.