Injector/Detector Plug Nuts, Restek
Catalog # RK21883
Supplier: Restek
Specifications
- Description:Injector/Detector Plug Nuts
- Cat. No.:RK21883
- Supplier no.:21883
Specifications
About this item
Use injector/detector plug nuts for a variety of purposes, including:
- Use to cap off an injector to isolate leaks.
- Use to cap off a detector for thermal cleaning.
- Use to check a detector or make-up gas flow rate.
- Use to cap off a detector and prevent hydrogen from accidentally diffusing into the oven from an unused detector base.