Deactivated Borosilicate Glass Wool, Restek
RK20789
: Restek
- Description:Deactivated Borosilicate Glass Wool
- Environmentally Preferable:
- Cat. No.:RK20789
- Supplier no.:20789
Deactivated borosilicate glass wool is more flexible than fused silica wool while still delivering exceptional performance.
- Deactivated borosilicate glass
- Flexible
Deactivated liners provide added insurance against possible sample adsorption.