WISP 48 Snap Seal Crimp-Top Vials, Restek
Supplier: Restek
Certificates
About this item
WISP 48 snap seal vials feature a crimp-top design and an extra-wide orifice size of 13 mm.
- Compatible with snap-seal caps
- Clear color
- Crimp-top format
Vials have a 4 ml capacity.