Big Mouth Step Vial Kits with Black Caps, Preassembled, Restek
Supplier: Restek
Certificates
About this item
10 mm big mouth step vial kits are preassembled for ready use.
- Caps attached to vials
- Black caps, red PTFE/silicone septa
- Screw-thread format
Vials have a 2 ml capacity.