Nylon Tube Brushes and Pipe Cleaner, Restek
Catalog # RK20108
Supplier: Restek
Specifications
- Bristle material:Nylon
- Description:Nylon Tube Brushes and Pipe Cleaner
- Cat. No.:RK20108
- Supplier no.:20108
Specifications
About this item
Use nylon tube brushes and pipe cleaners to remove small septum fragments and residue from dirty glass inlet liners.
- Brushes and pipe cleaners are available in different diameters
- Useful for fine cleaning operations
Brushes are 1/8, 3/16 and 1/4" in diameter; pipe cleaner is one foot (30 cm) long.