Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microscopes
Accessories for Boreal Digital Compound Microscopes
Accessories for Boreal Digital Compound Microscopes
Accessories for Boreal Digital Compound Microscopes
Catalog # 470021-964
Supplier:
LW Scientific
Expand image
Accessories for Boreal Digital Compound Microscopes
Catalog # 470021-964
Supplier:
LW Scientific
Supplier Number:
MSP-BLBH-1220
Specifications
Description:
Replacement Bulb 20W 12V Halogen
Cat. No.:
470021-964
Supplier no.:
MSP-BLBH-1220
Specifications
About this item
Product Details
Product Details