ContactUsLinkComponent
API® OF Medium Microbial Identification Kit, Biomerieux
API® OF Medium Microbial Identification Kit, Biomerieux
Catalog # 95060-850
Supplier:  bioMerieux
CAS Number:  
API® OF Medium Microbial Identification Kit, Biomerieux
Catalog # 95060-850
Supplier:  bioMerieux
Supplier Number:  50110
CAS Number:  
Specifications
  • Description:
    API® OF Medium Microbial Identification Kit
  • Size:
    10 x 5 mL
  • Environmentally Preferable:
  • Cat. No.:
    95060-850
  • Supplier no.:
    50110
Specifications
About this item
This medium is for use as a reagent with API® test strips for the determination of fermentation or oxidation metabolism.
  • Economical
  • Easy to use
  • Effective reagent
OF medium is recommended for the detection of oxidation or fermentation of carbohydrates by bacteria.