- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:10 μg
- Tag sequence:Fc Tag
- Storage Conditions:Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Protein Synonyms:c-Kit Ligand|Mast Cell Grow Factor|Stem Cell Factor|MGF|Kit Ligand
- Protein/Peptide Name:SCF
- Purity:>98% (SDS-PAGE)
- Sequence:26-187
- Formulation:Lyophilized from 0.2um-filtered solution in PBS.
- Cat. No.:75794-436
- Supplier no.:90-537
Stem cell factor (SCF), also known as cKit ligand (KL), mast cell growth factor (MGF) and steel factor (SLF), is a widely expressed 28-40 kDa type I transmembrane glycoprotein. It promotes the survival, differentiation and mobilization of multiple cell types including myeloid, erythroid, megakaryocytic, lymphoid, germ cell and melanocyte progenitors. SCF is a primary growth and activation factor for mast cells and eosinophils. Noncovalent dimers of transmembrane or soluble SCF interact with the receptor tyrosine kinase SCF R/cKit to trigger receptor dimerization and signaling. SCF assists in the recovery of cardiac function following myocardial infarction by increasing the number of cardiomyocytes and vascular channels.
- Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.