Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-21 mutant (from E. coli)
Human Recombinant IL-21 mutant (from E. coli)
Catalog # 75794-382
Supplier:  Prosci
CAS Number:  
Human Recombinant IL-21 mutant (from E. coli)
Catalog # 75794-382
Supplier:  Prosci
Supplier Number:  90-510
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 μg
  • Tag sequence:
    His Tag
  • Storage Conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein Synonyms:
    Interleukin-21
  • Protein/Peptide Name:
    IL-21 mutant
  • Purity:
    >90% (SDS-PAGE)
  • Sequence:
    32-162
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
  • Cat. No.:
    75794-382
  • Supplier no.:
    90-510

Specifications

About this item

Interleukin-21 (IL-21) is a key factor in the transition between innate and adaptive immune responses secreted by activated T cells. The IL-21 receptor (IL-21R) is expressed in lymphoid tissue, in particular by NK, B, T and dendritic cells, macrophages and endothelial cells. Recent evidence suggests that IL-21 plays a supportive role in the proliferation of T and B cells and influences the cytolytic activity of natural killer cells. IL-21 has been shown to up-regulate genes associated with innate immunity and to inhibit the differentiation of naive T helper cells. IL-21 specifically inhibits IFN-gamma production from developing TH1 cells and is preferentially expressed by TH2 cells. Furthermore IL-21 has been identified as a growth and survival factor for human myeloma cells. IL-21/IL-21R interactions have a unique role in sequentially activating both innate and adaptive immune responses against poorly immunogenic tumors, leading to tumor rejection that is perforin dependent but IFN-gamma independent.

  • Fusion-Tag: His Tag

This recombinant proteins is for research use only.