Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL18 (from E. coli)
Human Recombinant IL18 (from E. coli)
Catalog # 75794-374
Supplier:  Prosci
CAS Number:  
Human Recombinant IL18 (from E. coli)
Catalog # 75794-374
Supplier:  Prosci
Supplier Number:  90-506
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 μg
  • Tag sequence:
    His Tag
  • Storage Conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein Synonyms:
    Interferon-gamma-inducing Factor|IL1F4|Interleukin-18|IGIF|IL-1gamma
  • Protein/Peptide Name:
    IL18
  • Purity:
    >97% (SDS-PAGE)
  • Sequence:
    37-193
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
  • Cat. No.:
    75794-374
  • Supplier no.:
    90-506

Specifications

About this item

Interleukin-18 (IL-18) is a costimulatory factor for production of interferon-gamma (IFN-gamma) in response to toxic shock and shares functional similarities with IL-12. IL-18 is synthesized as a precursor 24kDa molecule without a signal peptide and must be cleaved to produce an active molecule. IL-1 converting enzyme (ICE; Caspase-1) cleaves pro-IL-18 at aspartic acid in the P1 position, producing the mature, bioactive peptide that is readily released from the cells. It is reported that IL-18 is produced from Kupffer cells, activated macrophages, keratinocytes, intestinal epithelial cells, osteoblasts, adrenal cortex cells and murine diencephalon. IFN-gamma is produced by activated T or NK cells and plays critical roles in the defense against microbiral pathogens. IFN-gamma activates macrophages and enhances NK activity and B cell maturation, proliferation and Ig secretion. IFN-gamma also induces expression of MHC class I and II antigens and inhibits osteoclast activation. IL-18 acts on T helper type-1 (Th1) T cells and in combination with IL-12 strongly induces them to produce IFN-gamma. Pleiotropic effects of IL-18 have also been reported, such as enhancement production of IFN-gamma and GM-CSF in peripheral blood mononuclear cells, production of Th1 cytokines, IL-2, GM-CSF, IFN-gamma in T cells and enhancement of Fas ligand expression by Th1 cells.

  • Fusion-Tag: His Tag

This recombinant proteins is for research use only.