Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant IL12 (from CHO Cells)
Catalog # 75794-368
Supplier:  Prosci
CAS Number:  
Mouse Recombinant IL12 (from CHO Cells)
Catalog # 75794-368
Supplier:  Prosci
Supplier Number:  90-503
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    25 μg
  • Tag sequence:
    Fc Tag
  • Storage Conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein Synonyms:
    NKSF|CLMF|Cytotoxic lymphocyte maturation factor|natural killer cell stimulatory factor|T-cell stimulating factor|TSF|IL12-p70
  • Protein/Peptide Name:
    IL12
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    23-215
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
  • Cat. No.:
    75794-368
  • Supplier no.:
    90-503

Specifications

About this item

Interleukin-12 (IL-12), also known as Natural Killer Cell Stimulatory Factor (NKSF) or Cytotoxic Lymphocyte Maturation Factor (CLMF), is a heterodimeric pleiotropic cytokine made up of a 40 kDa (p40) subunit and a 35 kDa (p35) subunit. IL-12 is produced by macrophages and B lymphocytes and has been shown to have multiple effects on T cells and Natural Killer (NK) cells. Some of these IL-12 activities include the induction of IFN-gamma and TNF in resting and activated T and NK cells; the enhancement of cytotoxic activity of resting NK and T cells, the stimulation of resting T cell proliferation in the presence of a comitogen; and the enhancement of NK cell proliferation. Current evidence indicates that IL-12 is a key mediator of cellular immunity and induces the differentiation of Th1 cells from precursor T helper cells. Based on its activities, it has been suggested that IL-12 may have therapeutic potential as a vaccine adjuvant that promotes cellular immunity and as an antitumor and anti viral agent.

  • Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.