Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CD279 (from CHO Cells)
Human Recombinant CD279 (from CHO Cells)
Catalog # 75794-360
Supplier:  Prosci
CAS Number:  
Human Recombinant CD279 (from CHO Cells)
Catalog # 75794-360
Supplier:  Prosci
Supplier Number:  90-447
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    100 μg
  • Tag sequence:
    Fc Tag
  • Storage Conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein Synonyms:
    PD-1|Programmed Cell Death Protein 1
  • Protein/Peptide Name:
    CD279
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    25-167
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
  • Cat. No.:
    75794-360
  • Supplier no.:
    90-447

Specifications

About this item

Programmed Death-1 (PD-1, CD279) is a type I transmembrane protein belonging to the CD28/CTLA-4 family of immunoreceptors that mediate signals for regulating immune responses. Members of the CD28/CTLA-4 family have been shown to either promote T cell activation (CD28 and ICOS) or downregulate T cell activation (CTLA-4 and PD-1). CD279 is expressed on activated T-cells, B-cells, myeloid cells and on a subset of thymocytes. In vitro, ligation of CD279 inhibits TCR-mediated T-cell proliferation and production of IL-1, IL-4, IL-10 and IFN-gamma. In addition, CD279 ligation also inhibits BCR mediated signaling. CD279 deficient mice have a defect in peripheral tolerance and spontaneously develop autoimmune diseases.

  • Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.