Human Recombinant CCL24 (from CHO cells)
Catalog # 75794-204
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:50 μg
- Tag sequence:Fc Tag
- Storage Conditions:Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Protein Synonyms:Small-inducible Cytokine A24|C-C Motif Chemokine 24|Myeloid Progenitor Inhibitory Factor 2|Eotaxin-2|MPIF-2|Eosinophil Chemotactic Protein 2|CKbeta6
- Protein/Peptide Name:CCL24
- Purity:>98% (SDS-PAGE)
- Sequence:27-119
- Formulation:Lyophilized from 0.2um-filtered solution in PBS.
- Cat. No.:75794-204
- Supplier no.:90-389
Specifications
About this item
CCL24 (eotaxin-2) has been shown to induce chemotaxis of eosinophils, basophils and resting T lymphocytes but not monocytes and activated T lymphocytes. CCL24 is a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. CCL24 interacts with chemokine receptor CCR3 to induce chemotaxis in eosinophils. Elevated levels of CCL24 has been seen in patients with aspirin-exacerbated respiratory disease (AERD), such as asthma.
- Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.