M. tuberculosis Recombinant NAD Synthetase (from E. coli)
Catalog # 75794-326
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:M. tuberculosis
- Size:50 μg
- Tag sequence:His Tag
- Storage Conditions:Stable for at least 6 months after receipt when stored at –80 °C.
- Enzyme Name:NAD Synthetase
- Enzyme Activity:~0.1 U/mg protein. (Ammonium or glutamine as nitrogen source). One unit is defined as the amount of enzyme that synthesizes 1umol NAD per min.
- Enzyme Synonyms:NADE, Glutamine-dependent NAD(+) Synthetase, EC 6.3.5.1
- Purity:>98% (SDS-PAGE)
- Formulation:Liquid, In 20 mM TRIS-HCl, pH 7.8.
- Cat. No.:75794-326
- Supplier no.:90-366
Specifications
About this item
NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalyzing the final step of both pathways
This recombinant proteins is for research use only.
Fusion-Tag: His Tag
Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It can use both glutamine or ammonia as a nitrogen source. Catalytic activity: ATP + deamido-NAD+ + L-glutamine + H2O = AMP + diphosphate + NAD+ + L-glutamate