Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant PARP1 E988K Mutant Control (from Insect Cells (Sf21))
Human Recombinant PARP1 E988K Mutant Control (from Insect Cells (Sf21))
Catalog # 75794-152
Supplier:  Prosci
CAS Number:  
Human Recombinant PARP1 E988K Mutant Control (from Insect Cells (Sf21))
Catalog # 75794-152
Supplier:  Prosci
Supplier Number:  90-343
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells (Sf21)
  • Species:
    Human
  • Size:
    10 μg
  • Tag sequence:
    His Tag
  • Storage Conditions:
    Stable for at least 6 months after receipt when stored at −80 °C.
  • Biological Activity:
    0.5% of wild type PARP-1.
  • Protein Synonyms:
    ARTD1
  • Protein/Peptide Name:
    PARP1 E988K Mutant Control
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Liquid, In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride and 50mM imidazole, 0.2% NP-40 and 10% glycerol.
  • Cat. No.:
    75794-152
  • Supplier no.:
    90-343

Specifications

About this item

PARP-1 (ARTD1) is involved in the base excision repair (BER) pathway, by catalyzing the poly(ADP-ribosyl)ation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signaling pathway leading to the reparation of DNA strand breaks. PARP-1 positively regulates the transcription of MTUS1 and negatively regulates the transcription of MTUS2/TIP150. It forms a complex with EEF1A1 and TXK that acts as a T-helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production. PARP-1 (E998K mutant) is an inactive form of PARP-1 which can be used as a control compound.

  • Fusion-Tag: His Tag

This recombinant proteins is for research use only.