Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant FGF1 (from E. coli)
Mouse Recombinant FGF1 (from E. coli)
Catalog # 75794-124
Supplier:  Prosci
CAS Number:  
Mouse Recombinant FGF1 (from E. coli)
Catalog # 75794-124
Supplier:  Prosci
Supplier Number:  90-329
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    10 μg
  • Tag sequence:
    Tag Free
  • Storage Conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Protein Synonyms:
    HBGF-1|Heparin-binding Growth Factor 1|Endothelial Cell Growth Factor|Acidic Fibroblast Growth Factor|Fibroblast Growth Factor 1|ECGF
  • Protein/Peptide Name:
    FGF1
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    16-155
  • Formulation:
    Lyophilized, In PBS containing 0.2 mM DTT.
  • Cat. No.:
    75794-124
  • Supplier no.:
    90-329

Specifications

About this item

Fibroblast growth factors (FGFs) constitute a family of heparin-binding polypeptides involved in the regulation of biological responses such as growth, differentiation and angiogenesis. The biological effects of FGFs are mediated by four structurally related receptor tyrosine kinases denoted FGFR1, FGFR2, FGFR3 and FGFR4. FGF-1 [FGF-acidic; ECGF; HBGF-1] is a powerful mitogen of cells of mesodermal, ectodermal and endodermal origin. FGF-1 association with heparan sulfate is a prerequisite for activation of FGF receptors. FGF-1 plays a role in various stages of development and morphogenesis as well as in angiogenesis and wound healing processes. Recent data indicate a role of FGF-1 in inflammation and obesity. FGF-1 is selectively induced in fat cells by high-fat diet feeding and established the PPARgamma-FGF-1 axis as a critical pathway that regulates adipose tissue remodeling.

  • Fusion-Tag: Tag Free

This recombinant proteins is for research use only.