Human Recombinant Angiopoietin 2 (from HEK293 cells)
Catalog # 75793-824
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:10 μg
- Tag sequence:DDDDK Tag
- Storage Conditions:Stable for at least 6 months after receipt when stored at −20 °C.
- Protein Synonyms:ANGPT2|ANG2|Angiopoietin-2|AGPT2
- Protein/Peptide Name:Angiopoietin 2
- Purity:>95% (SDS-PAGE)
- Sequence:19-496
- Formulation:Lyophilized, Contains PBS with trehalose and Brij35.
- Cat. No.:75793-824
- Supplier no.:90-311
Specifications
About this item
Angiopoietin-1 (Ang-1) and Angiopoietin-2 (Ang-2) are closely related secreted ligands which bind with similar affinity to Tie-2. Tie-2 and angiopoietins have been shown to play critical roles in embryogenic angiogenesis and in maintaining the integrity of the adult vasculature. Ang-1 activates Tie-2 signaling on endothelial cells to promote chemotaxis, cell survival, cell sprouting, vessel growth and stabilization. Ang-2 has been identified as a secreted protein ligand of Tie-2 and has alternatively been reported to be an antagonist for Ang-1 induced Tie-2 signaling as well as an agonist for Tie-2 signaling, depending on the cell context.
- Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.