Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant CD40L multimeric (from CHO Cells)
Catalog # 75794-084
Supplier:  Prosci
CAS Number:  
Human Recombinant CD40L multimeric (from CHO Cells)
Catalog # 75794-084
Supplier:  Prosci
Supplier Number:  90-289
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    10 μg
  • Tag sequence:
    DDDDK Tag
  • Storage Conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Biological Activity:
    Induces B cell activation (as demonstrated by dose-dependent upregulation of CD86).
  • Protein Synonyms:
    ADIPOQ-CD40L|ACRP30headless:CD154|ACRP30headless:TNFSF5|ACRP30headless:CD40L
  • Protein/Peptide Name:
    CD40L multimeric
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    18-111
  • Formulation:
    Lyophilized, Contains PBS.
  • Cat. No.:
    75794-084
  • Supplier no.:
    90-289

Specifications

About this item

MultimericCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo. It provides a simple and equally potent alternative to CD40L+enhancer combinations. MultimericCD40L™ has been shown to suppress alum-induced IL-1beta release and caspase-1 activation in a dose-, CD40- and time dependent manner, without affecting BMDM (Bone marrow-derived macrophages) viability. It also effectively suppressed the inflammasome function triggered by NLRP3 activators. The secretion of caspase-1 independent inflammatory mediators has been shown to be unaltered or even enhanced.

  • Fusion-Tag: DDDDK Tag

This recombinant proteins is for research use only.