Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant APRIL multimeric (from HEK293 cells)
Catalog # 75794-052
Supplier:  Prosci
CAS Number:  
Human Recombinant APRIL multimeric (from HEK293 cells)
Catalog # 75794-052
Supplier:  Prosci
Supplier Number:  90-273
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    10 μg
  • Tag sequence:
    DDDDK Tag
  • Storage Conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Biological Activity:
    Stimulates B cell proliferation.
  • Protein Synonyms:
    ACRP30headless:TNFSF13|ACRP30headless:APRIL|ACRP30headless:A-proliferation-inducing Ligand:|ACRP30headless:CD256
  • Protein/Peptide Name:
    APRIL multimeric
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    18-110
  • Formulation:
    Lyophilized, Contains PBS + 0.5% Trehalose.
  • Cat. No.:
    75794-052
  • Supplier no.:
    90-273

Specifications

About this item

APRIL is a cytokine that belongs to the TNF superfamily and binds to TACI and BCMA. It is implicated in the regulation of tumor cell growth, is involved in monocyte/macrophage-mediated immunological processes and functions as an important survival factor for plasmablasts and bone marrow plasma cells. MultimericAPRIL™ is a high activity construct in which two trimeric APRIL ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively stimulates proliferation B cell. A basic amino acid sequence (QKQKKQ) close to the N-terminus of APRIL is required for binding to negatively charged sulfated glycosaminoglycan side chains of proteoglycans. Proteoglycans mediate binding of APRIL to tumor cells as well as primary lymphoid cells.

  • Fusion-Tag: DDDDK Tag

This recombinant proteins is for research use only.