Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant ANGPTL4 (from COS-7 cells)
Mouse Recombinant ANGPTL4 (from COS-7 cells)
Catalog # 75793-654
Supplier:  Prosci
CAS Number:  
Mouse Recombinant ANGPTL4 (from COS-7 cells)
Catalog # 75793-654
Supplier:  Prosci
Supplier Number:  90-074
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    COS-7 cells
  • Species:
    Mouse
  • Size:
    10 μg
  • Tag sequence:
    DDDDK Tag
  • Storage Conditions:
    Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological Activity:
    Activates ERK phosphorylation in THP-1 cells.
  • Protein Synonyms:
    PSEC0166|Angiopoietin-related protein 4|ANGPTL4|Hepatic fibrinogen/angiopoietin-related protein|ARP4|HFARP|PP1158|FIAF|PGAR|UNQ171/PRO197
  • Protein/Peptide Name:
    ANGPTL4
  • Purity:
    >90% (SDS-PAGE)
  • Sequence:
    1-410
  • Formulation:
    0.2um-filtered solution in PBS.
  • Cat. No.:
    75793-654
  • Supplier no.:
    90-074

Specifications

About this item

ANGPTL4 (Angiopoietin-like protein 4) mainly expressed in endothelial cells (hypoxia-induced). Regulates angiogenesis and modulates tumorigenesis and directly regulates lipid, glucose, and energy metabolism. Inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage. ANGPTL4 is a protein consisting of an N-terminal coiled-coil domain and a C-terminal fibrinogen-like domain (FLD). Both domains have distinct biological functions. The coiled-coil domain is responsible for the inhibitory effects on lipoprotein lipase (LPL) converting the active form of LPL into an inactive form, and the FLD domain mediates its antiangiogenic functions. The coiled coil and the FLD domains are separated by a short linker that can be cleaved after secretion. ANGPTL4 appears on the cell surface as the full-length form, where it can be released by heparin treatment. ANGPTL4 protein is then proteolytically cleaved by proprotein convertases (PCs), including furin, PC5/6, paired basic amino acid-cleaving enzyme 4, and PC7.

  • Fusion-Tag: DDDDK Tag

This recombinant proteins is for research use only.